Learn More
Waters Corp Waters Corporation HPLC Column, Symmetry300 C18, 3.5 μm, 4.6 x 150 mm
Supplier:  Waters Corp 186000197
Encompass_Preferred
These Symmetry300 C18 columns are wide-pore, reversed-phase chromatography columns, designed for superior performance in separating hydrophilic and hydrophobic peptides, neutral, and highly basic peptides. They deliver high peptide recovery and are quality control tested with peptide samples to ensure exceptional batch-to-batch reproducibility, suitable for applications requiring high or low mass loads.
  • Quality control tested with peptides.
  • Wide pore (300 Å).
  • Reversed-phase columns.
  • Provides high recoveries of peptides and proteins.
  • Delivers maximum reproducibility batch-to-batch and column-to-column.
  • Provides superior peak shape.
  • Manufactured using high purity silica and tightly controlled manufacturing processes.
Catalog No. 50-818-728