Learn More
Waters Corp Waters Corporation Column, C18, 5 μm, 300 Å, 3.9 mm × 150 mm
Supplier: Waters Corp WAT106154
These columns primarily support legacy methods and set the standard for HPLC drug assays, providing confidence in the long-term compliance of HPLC methods. Manufactured with tight specifications and high purity silica, they offer consistent results and outstanding reproducibility for established analytical methods. They are designed for long lifetimes, delivering superior peak shape across a wide pH range.
- Chemistry: C18; Separation Mode: Reversed Phase.
- Particle Substrate: Spherical Silica; Particle Size: 5 μm.
- Pore Size: 300 Å.
- Dimensions: 3.9 mm Inner Diameter × 150 mm Length.
- pH Range: 2 to 8.
- Maximum Pressure: 6000 psi (415 Bar).
- Endcapped with low silanol activity.
- Carbon Load: 8.5%.
- Designed for over 10,000 injections with minimal performance change.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.