Learn More
Waters Corp WATERS CORPORATION Column, C4, 300 Å, 5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186000288
Encompass_Preferred
These HPLC columns offer excellent peak shape, selectivity, and reproducibility for sensitive analysis of large peptides and proteins in the pharmaceutical industry.
  • Supports legacy HPLC methods and long-term compliance.
  • Sets standard for drug assays; superior peak shape for active peptides.
  • Effective for diverse peptides: hydrophilic, hydrophobic, neutral, basic.
  • Performs well under high and low mass conditions.
  • Recommended for mass spectrometry with low TFA or 0.1% formic acid.
  • Optimal for validation compliance.
  • Delivers sharpest peaks for biomolecules and complex tryptic maps.
  • Ultrapure silica with low silanol activity.
  • High efficiency packing for improved precision and quantitation.
Catalog No. 50-818-731