Learn More
Waters Corp Waters Column, C4, 300 Å, 3.5 μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186000282
Encompass_Preferred
This C4 column supports legacy HPLC methods, ensuring long-term compliance with consistent peak shape, selectivity, and reproducibility. Optimized for separating physiologically active peptides, it performs efficiently in varied mass situations, ideal for mass spectrometry applications using low TFA or 0.1% formic acid mobile phases.
  • Supports legacy HPLC methods, ensuring long-term compliance.
  • Delivers consistent peak shape, selectivity, and reproducibility.
  • Excels in separating physiologically active peptides (hydrophilic, hydrophobic, neutral, highly basic).
  • Provides optimal performance in both high and low mass applications.
  • Features high purity and very low silanol activity from ultrapure silica.
  • Packed for maximum efficiency and reproducibility, improving precision for impurity and stability tests.
Catalog No. 50-780-169