Learn More
Waters Corp Waters Column, C4, 300 Å, 5 μm, 4.6 mm × 50 mm
Supplier:  Waters Corp 186000287
Encompass_Preferred
This HPLC reversed-phase column with C4 chemistry is designed for precise drug testing and separation of large peptides and proteins. It offers excellent peak shape, selectivity, and reproducibility, using ultrapure silica for high purity and low silanol activity, making it suitable for demanding analytical applications, including mass spectrometry.
  • Supports legacy HPLC methods and ensures long-term compliance.
  • Maintains excellent peak shape, selectivity, and reproducibility.
  • Designed for separating large peptides, proteins, and various peptide types.
  • Compatible with mass spectrometry, recommending low TFA mobile phases.
  • Utilizes ultrapure silica for high purity and low silanol activity.
  • Packed for optimal efficiency, reproducibility, and precise quantitation.
  • Offers a wide operating pH range of 2 to 8.
  • Lifetime can be extended with a compatible VanGuard Cartridge.
Catalog No. 50-780-174