Learn More
Waters Corp Waters Corporation Column, C18, 100Å, 7 μm, 7.8 x 300 mm
Supplier: Waters Corp WAT066235
These C18 Prep Columns are engineered to meet the demanding requirements of HPLC drug assays, ensuring long-term compliance and consistent performance. They provide exceptional peak shape, selectivity, and reliable reproducibility, thereby avoiding typical performance compromises.
- C18 chemistry
- Reversed phase separation mode
- Silica particle substrate
- pH range of 2 to 8
- Temperature limits of 45 °C
- Maximum pressure of 6000 psi (415 Bar)
- Endcapped
- Low silanol activity
- Spherical particle shape
- 7 μm particle size
- 100 Å pore size
- Prep column format
- Suitable for HPLC preparative systems
- Supports LC/MS and LC techniques
- USP classification L1
- 7.8 mm inner diameter
- 300 mm length
- 19% carbon load
- Units per package: 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.