Learn More
Waters Corp Waters Corporation Prep Column, C8, 5 μm, 7.8 mm, 50 mm
Supplier: Waters Corp 186000214
Symmetry columns continue to set the standard for HPLC drug assays, providing confidence in the long-term compliance of HPLC methods. They offer excellent peak shape, selectivity, column-to-column and batch-to-batch reproducibility, and other critical performance characteristics. Available in a variety of particle sizes, chemistries, and dimensions.
- C8 chemistry.
- Reversed phase separation mode.
- Particle substrate: Silica.
- pH range 2 to 8.
- Temperature limits: 45°C.
- Maximum pressure: 6000 psi (415 Bar).
- Particle size: 5 μm.
- Pore size: 100 Å.
- Inner diameter: 7.8 mm.
- Length: 50 mm.
- Endcapped.
- Low silanol activity.
- Spherical particle shape.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.