Learn More
Waters Corp Waters Symmetry C8 Prep Column, 7 μm, 19 mm X 150 mm, 100 Å
Supplier: Waters Corp WAT066228
These columns are designed to set the standard for HPLC drug assays, ensuring long-term compliance for HPLC methods. They aim to provide consistent peak shape, selectivity, and reproducibility across columns and batches. These columns are available in various particle sizes, chemistries, and dimensions.
- Features C8 chemistry for reversed-phase separation with a silica substrate.
- Offers a pH range from 2 to 8 and temperature limits up to 45°C.
- Withstands a maximum pressure of 6000 psi (415 Bar).
- Endcapped design ensures low silanol activity and spherical particle shape.
- Particle size of 7 μm and a pore size of 100 Å.
- Designed as a preparative column for HPLC, suitable for LC/MS and LC techniques.
- Conforms to USP classification L7.
- Measures 19 mm inner diameter and 150 mm length with 12% carbon load.
- Includes one unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.