Learn More
Waters Corp Waters HPLC Column, C18, 3.5 μm, 4.6 mm x 150 mm
Supplier: Waters Corp 186000180
These reversed-phase columns feature innovative embedded polar group technology, which shields the silica’s residual silanol surface from highly basic analytes. This technology significantly improves peak shape and resolution, making these columns ideal for applications requiring low organic, highly aqueous mobile phase conditions.
- Contains embedded polar group technology that “shields” the silica’s residual silanol surface from highly basic analytes.
- Reduced silanol activity significantly improves peak shape and resolution.
- Delivers USP Tailing Factors of less than 1.2 across a pH range of pH 2 to pH 8.
- Ideal for applications that require low organic, highly aqueous mobile phase conditions.
- Ensures reproducibility across high performance liquid chromatography applications.