Learn More
Waters Corp Symmetry Shield RP18 Column, 100Å, 3.5 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186000699
Encompass_Preferred
These reversed-phase columns contain Waters patented embedded polar group technology that “shields” the silica’ residual silanol surface from highly basic analytes. The reduced silanol activity for the SymmetryShield RP18 column significantly improves peak shape and resolution.
Catalog No. 50-780-465