Learn More
Waters Corp Waters Symmetry Shield RP18 Column, 100Å, 3.5 μm, 3 mm X 50 mm
Supplier:  Waters Corp 186002614
Encompass_Preferred
These reversed-phase columns contain patented embedded polar group technology that “shields” the silica’s residual silanol surface from highly basic analytes. The reduced silanol activity significantly improves peak shape and resolution.
  • C18 chemistry
  • Reversed-phase separation mode
  • Silica particle substrate
  • pH range of 2–8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Shield RP18 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • Column format
  • Surface area: 335
  • HPLC system
  • USP classification L1
  • 17% carbon load
  • 1 unit per package
Catalog No. 50-781-243