Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Symmetry Shield RP18 Column, 3 x 150 mm, 5 μm

Supplier:  Waters Corp 186000692

Encompass_Preferred

These reversed-phase columns contain embedded polar group technology that “shields” the silica's residual silanol surface from highly basic analytes. The reduced silanol activity significantly improves peak shape and resolution.

  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped design
  • Shield RP18 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100 Å
  • HPLC system compatible
  • USP L1 classification
  • 3 mm inner diameter
  • 150 mm length
  • 17% carbon load
  • One unit per package

Catalog No. 50-780-458


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.