Learn More
Waters Corp Waters Column, 100Å, 5 μm, 3 mm × 250 mm
Supplier:  Waters Corp 186000693
Encompass_Preferred
These reversed-phase columns incorporate patented embedded polar group technology, effectively shielding the silica's residual silanol surface from highly basic analytes to improve peak shape and resolution.
  • Significantly improves peak shape and resolution for basic analytes by shielding residual silanol surfaces.
  • Provides improved peak form across a wide pH range, allowing optimization at neutral or low pH.
  • Ensures durability and cost-effectiveness through hydrolytic and mechanical stability of packing material.
  • Offers remarkable reproducibility due to rigorous batch testing and narrow specification ranges.
  • Features the industry's smallest ligand surface coverage, minimizing resolution shifts.
  • Manufactured in a cGMP, ISO 9001 certified facility using ultra-pure reagents.
Catalog No. 50-780-459