Learn More
Waters Corp Waters Chromatography Column, C18, 5 μm, 2.1 mm X 150 mm
Supplier:  Waters Corp 186000111
Encompass_Preferred
These reversed-phase columns feature patented embedded polar group technology, which “shields” the silica’s residual silanol surface from highly basic analytes, improving peak shape and resolution. The column provides improved peak form and resolution across a broad pH range and retains basic chemicals less than conventional C18 and C8 columns.
  • Durable packing material and mechanical stability contribute to longevity, reducing lab equipment costs.
  • Offers a wide pH range, with high bonding density and low silanol activity, maintaining peak shape at neutral or low pH.
  • Outstanding reproducibility due to tight specification ranges and narrowest ligand surface coverage, minimizing resolution shifts batch-to-batch.
Catalog No. 50-780-071