Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HPLC Column, SymmetryShield RP18, 3.5 μm, 3.9 x 20 mm

Supplier:  Waters Corp 186002084

Encompass_Preferred

This product is an HPLC Column, specifically the SymmetryShield RP18 with a particle size of 3.5 μm and dimensions of 3.9 x 20 mm. It is suitable for various liquid chromatography applications.

  • Phase type: RP18 (SymmetryShield)
  • Particle size: 3.5 μm
  • Dimensions: 3.9 x 20 mm

Catalog No. 50-780-940


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.