Learn More
Waters Corp Waters Symmetry Shield RP8 Column, 100 Å, 5 μm
Supplier:  Waters Corp WAT094243
Encompass_Preferred
These reversed-phase columns incorporate patented embedded polar group technology, which shields the silica’s residual silanol surface from highly basic analytes. This reduces silanol activity, improving peak shape and resolution. Ideal for very aqueous mobile phases, the embedded polar group inhibits pore dewetting, ensuring analyte retention and eliminating lengthy re-wetting processes.
  • Improved peak shape and resolution.
  • Inhibits pore dewetting for aqueous mobile phases.
  • High reproducibility with tight ligand surface coverage.
  • Extended column lifetime.
  • Excellent method development tool.
  • Superior peak shape across pH range.
  • Increased productivity and faster method development.
Catalog No. 50-785-997