Learn More
Biotium SynaptoGreen™C3
Supplier:  Biotium 70023
Encompass_Preferred
SynaptoGreen™ C3 is similar to SynaptoGreen™ C4, except that the lipophilic tail is one carbon shorter and also the hydrophilic end is a trimethylammonium group. Thus SynaptoGreen™ C3 is slightly more water soluble than SynaptoGreen™ C4. Properties: Ex/Em: 510/625 nm (in MeOH); 480/598 nm (in membranes); Red solid soluble in water; C25H39Br2N3; MW: 542
Catalog No. 50-196-4753