Learn More
Medchemexpress LLC Syndecan-4 Human 50ug
SDP
Supplier:  Medchemexpress LLC HYP7192750UG
Encompass_Preferred
Recombinant human Syndecan-4 is supplied as a purified, tag-free protein expressed in E. coli and provided lyophilized for use in biochemical and functional assays. The material is validated by reducing SDS-PAGE (>95% purity) and demonstrates binding activity to basic FGF in ELISA (ED50 = 7.83 ng/mL).
  • High purity for reproducible biochemical and binding assays.
  • Tag-free format preserves native protein properties.
  • Validated functional activity in ELISA with a defined ED50.
  • Lyophilized form for improved stability and storage.
  • Suitable for ELISA, binding studies, and assay development.
Catalog No. 50-001-96368