Learn More
ILS INFRARED LABORATORY SYSTEMS LLC Synermed IR170-R MAGNESIUM
Supplier:  ILS INFRARED LABORATORY SYSTEMS LLC IR170R
Encompass_Preferred
The Synermed magnesium is detected in the infrared region of the spectrum thus avoiding the absorbance of hemoglobin and minimizing the absorbance of lipemia. Except in geriatric populations, most pathologic magnesium values are below normal. Consequently, signal levels [absorbance changes] are very low. Low levels of serum background noise is very important to avoid chromatic interference, especially in the important pathologic low levels.Single Liquid-Stable Ready-to-Use ReagentMinimal Interference from Serum Bilirubin, Hemoglobin and LipemiaExtended "On-Board" Stability
Catalog No. NC3877143