Learn More
ILS INFRARED LABORATORY SYSTEMS LLC Synermed VI400-R AMYLASE
Supplier:  ILS INFRARED LABORATORY SYSTEMS LLC VI400R
Encompass_Preferred
Liquid-StableSingle Ready-to-Use ReagentImproved SensitivityMinimal Bilirubin InterferenceNo Maltase Interference - Usable with Veterinary Samples
Catalog No. NC3877170