Learn More
Medchemexpress LLC SYNGAP1 Human His 20ug

Supplier: Medchemexpress LLC HYP7161920UG
Recombinant human SYNGAP1 protein fragment (residues M1161-H1343) expressed in E. coli with an N-terminal His tag and supplied as a lyophilized powder. The fragment (~28 kDa) is a postsynaptic density component involved in Ras-cAMP and NMDAR-mediated signaling; it is provided at high purity for research applications and should be stored and handled according to product instructions.
- Partial sequence corresponding to residues M1161-H1343.
- Approximate molecular weight 28 kDa.
- Expressed in E. coli with an N-terminal His tag.
- Purity greater than 90% by reducing SDS-PAGE.
- Supplied lyophilized from 10 mM Tris-HCl, 1 mM EDTA, pH 8.0.
- Size/quantity 20 μg.
- Intended for scientific research use only.
- Recommended to aliquot and freeze at -20°C or -80°C for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.