Learn More
Bachem C-Peptide (human) Acetate
SDP
Supplier:  Bachem 4068086.0500
Encompass_Preferred
C-Peptide (human) Acetate, H-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-OH, 33017-11-7, C129H211N35O48, Mr 3020.30, 0.5mg.
Catalog No. 50-194-6877