Learn More
Cayman Chemical SynthetIc AlIzarIn 50g
SDP
Supplier:  Cayman Chemical 1167650 G
Encompass_Preferred
Inhibitor of P450 isoforms CYP1A1, CYP1A2, and CYP1B1; antioxidant activity in rats; reduces hepatic content of reactive substances and serum aminotransferases in poisoned rats; a small molecule red dye
Catalog No. 50-295-1979