Learn More
ATCC Synthetic Astrovirus; RNA
Supplier:  ATCC VR3238SD
Encompass
Quantitative synthetic RNA for Astrovirus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes fragments from the ORF1a, ORF1b, ORF2, and 3' UTR regions.
Catalog No. 50-238-3792