Learn More
Cayman Chemical SynthetIc AvIcularIn 1mg
SDP
Supplier:  Cayman Chemical 339181 MG
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; inhibits FASN and aldose reductase (IC50s = 6.15 and 19.05 µM for the chicken liver and human enzymes, respectively); scavenges DPPH and superoxide radicals, as well as inhibits copper-induced oxidation of LDL in cell-free assays (IC50s = 64.3, 6, and 3.8 µM, respectively); inhibits LPS-induced increases in NO and PGE2 production in RAW 264.7 macrophages at 30-300 µM; inhibits the proliferation, migration, and invasion of, as well as induces apoptosis and cell cycle arrest at the G0/G1 phase in, Huh7 cells at 100 UG/ml
Catalog No. 50-294-9915