Learn More
Cayman Chemical SynthetIc Bergaptol 1mg
SDP
Supplier:  Cayman Chemical 387421 MG
Encompass_Preferred
A furanocoumarin with diverse biological activities; inhibits CYP3A4 (IC50 = 24.92 µM); scavenges DPPH and ABTS radicals in cell-free assays (EC50s = 26.66 and 13.55 UG/ml, respectively); reduces the replication of L. donovani, T. b. brucei, and T. cruzi in L6 myoblasts (IC50s = 2.5, 22.1, and 68 UG/ml, respectively); induces cell cycle arrest in the S phase and apoptosis in MCF-7 cells in a concentration-dependent manner; decreases the proliferation of HeLa and HepG2 cells (IC50s = 58.57 and 68.42 µM, respectively)
Catalog No. 50-291-7468