Learn More
Cayman Chemical SynthetIc C82 1mg
SDP
Supplier:  Cayman Chemical 335631 MG
Encompass_Preferred
An Mtb CdnP inhibitor (IC50 = 17.5 µM); selective for Mtb CdnP over Yybt, RocR, and GBS CdnP, ENPP1, and poxin at 200 µM
Catalog No. 50-294-8973