Learn More
Cayman Chemical SynthetIc CerberIn 2.5mg
SDP
Supplier:  Cayman Chemical 355822.5 MG
Encompass_Preferred
A cardiac glycoside with cytotoxic and cardiac modulatory activities; cytotoxic to KB and NCI H187 cancer cells (IC50s = 1.92 and 1.24 UG/ml); inhibits Na+/K+-ATPase, increasing intracellular sodium levels and action potential duration in cardiac cells
Catalog No. 50-291-3444