Learn More
Cayman Chemical SynthetIc cIs-ZeatIn 10mg
SDP
Supplier:  Cayman Chemical 3554910 MG
Encompass_Preferred
A cytokinin plant growth regulator; induces callus growth in isolated N. tabacum callus tissue from 0.1-1 µM
Catalog No. 50-291-3615