Learn More
Cayman Chemical SynthetIc DIeugenol 10mg
SDP
Supplier:  Cayman Chemical 2755710 MG
Encompass_Preferred
A neolignan with antioxidative and antiprotozoal activities; inhibits TBARS formation and scavenges superoxide anions, but not hydroxyl radicals, in cell-free assays; has anti-trypanosomal activity against T. cruzi amastigotes and trypomastigotes (IC50s = 15.1 and 11.5 µM, respectively) but is cytotoxic to NCTC L-929 fibroblasts (CC50 = 58.2 µM); disrupts the integrity of the T. cruzi trypomastigote plasma membrane but does not induce ROS production in trypomastigotes or LPS-stimulated and unstimulated isolated mouse peritoneal macrophages
Catalog No. 50-293-8630