Learn More
ATCC Synthetic Hepatitis B; virus DNA
Supplier:  ATCC VR3232SD
Encompass
Quantitative synthetic DNA for Hepatitis B virus can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. This preparation includes fragments from the highly conserved precore, core, P, S, and X regions.
Catalog No. 50-238-2403