Learn More
Cayman Chemical SynthetIc ID-8 10mg
SDP
Supplier:  Cayman Chemical 1522210 MG
Encompass_Preferred
A cell culture supplement that can sustain self-renewal and pluripotency of mouse embryonic stem cells at 10 μM for > 30 days
Catalog No. 50-297-2337