Learn More
Cayman Chemical SynthetIc Lupulon 1mg
SDP
Supplier:  Cayman Chemical 284811 MG
Encompass_Preferred
A beta-acid; active against B. subtilis and S. aureus (MICs = 1 and 1.2 UG/ml, respectively), and T. b. brucei and L. m. mexicana (IC50s = 0.9 and 4.7 UG/ml, respectively); scavenges DPPH radicals in a cell-free assay and inhibits lipid peroxidation in rat brain homogenates (IC50s = 25 and 39 µM, respectively); reduces proliferation, cell migration, and capillary tube formation in HUVECs at 2.5 to 50 UG/ml; lupulone (40 UG/ml) activates the extrinsic apoptotic death pathway in SW480 and SW620 colon cancer cells.
Catalog No. 50-293-8285