Learn More
Cayman Chemical SynthetIc MelatonIn 10g
SDP
Supplier:  Cayman Chemical 1442710 G
Encompass_Preferred
An indoleamine neurohormone whose levels vary in a daily cycle, thereby allowing the entrainment of the circadian rhythms of several biological functions in animals, plants, and microbes; acts as a powerful antioxidant that protects lipids, proteins, and DNA against oxidative damage
Catalog No. 50-297-2441