Learn More
ATCC Synthetic Mycoplasma; genitalium DNA
Supplier:  ATCC BAA2641SD
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day to day test variation and lot to lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. Preparation includes fragments from the 16S gene, mgpA, and gap.
Catalog No. 50-238-4036