Learn More
Synthetic Perspiration Reagent ANSI/BHMA A156.18-2020, Reagents
Synthetic Perspiration Reagent ANSI/BHMA A156.18-2020, Reagent, for general laboratory use. Recommended Storage: 2to 8°C (36°C to 46°F) Manufactured in ISO 9001 facility.
$194.80 - $327.00
Chemical Identifiers
| CAS | 7732-18-5 , 7647-14-5 , 64-19-7 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Artificial sweat |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-333-494
|
Reagents Holdings Llc
CS5001001A |
1 L |
Each for $327.00
|
|
|||||
|
01-333-493
|
Reagents Holdings Llc
CS500100500A |
500 mL |
Each for $194.80
|
|
|||||
Chemical Identifiers
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Colorless | |
| 100°C | |
| Artificial sweat | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <7 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.