Learn More
Synthetic Perspiration Reagent ANSI/BHMA A156.18-2020, Reagents
Synthetic Perspiration Reagent ANSI/BHMA A156.18-2020, Reagent, for general laboratory use. Recommended Storage: 2to 8°C (36°C to 46°F) Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CS500100500A
| Quantity | 500 mL |
|---|
Chemical Identifiers
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Colorless | |
| 100°C | |
| Artificial sweat | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <7 | |
| Non-sterile | |
| Synthetic Perspiration Reagent ANSI/BHMA A156.18-2020. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.