Learn More
Cayman Chemical SynthetIc PF-1022A 1mg
SDP
Supplier:  Cayman Chemical 297001 MG
Encompass_Preferred
A cyclodepsipeptide with anthelmintic activity; oral administration induces worm excretion in a chicken model of A. galli infection from 0.5-2 mg/kg; induces parasite clearance in jird models of H. contortus, O. ostertagi, or T. colubriformis infection (ED95s = 0.11, 0.33, and 0.11 mg/animal)
Catalog No. 50-299-1637