Learn More
Cayman Chemical SynthetIc PImodIvIr 5mg
SDP
Supplier:  Cayman Chemical 293215 MG
Encompass_Preferred
An inhibitor of influenza virus PB2 (KD = ~1.6 µM) and inhibits the activity of Axl and CaMKIIβ by >50% in a panel of 65 human and rat kinases; decreases the replication of seven adamantine- and neuraminidase inhibitor-resistant strains of influenza virus A (EC50s = 50s = 0.58, 0.64, and 0.89, respectively, in a cell-based assay); increases survival in a mouse model of intranasal influenza A infection at 1, 3, and 10 mg/kg twice per day
Catalog No. 50-299-2607