Learn More
Cayman Chemical SynthetIc PInosylvIn 1mg
SDP
Supplier:  Cayman Chemical 276121 MG
Encompass_Preferred
A stilbene with diverse biological activities; activates SIRT1 and induces glucose uptake in isolated rat L6 skeletal muscle myotubes; reduces radial growth in a panel of 28 plant pathogenic fungi at 100 μg/ml; reduces expression of MMP-2, MMP-9, and membrane type 1-MMP in and inhibits migration of HT-1080 cells; reduces the number of tumor nodules and lung tumor weight in a CT26 mouse xenograft model of metastatic colon cancer at 10 mg/kg; decreases hind paw volume and MPO activity in a rat model of adjuvant-induced arthritis; exhibits plant antifeedant activity against L. americanus
Catalog No. 50-294-0491