Learn More
Cayman Chemical SynthetIc SpongosIn 10mg
SDP
Supplier:  Cayman Chemical 4183210 MG
Encompass_Preferred
A nucleoside with diverse biological activities; antagonist of adenosine A1, A2A, and A3 receptors (Kis = 155, 970, and 156 nM, respectively, in CHO cells expressing the human receptors); inhibits collagen-induced platelet aggregation in isolated human whole blood and platelet-rich plasma at 100 µM; decreases contractile force and frequency in spontaneously contracting isolated guinea pig atrial muscles (EC50s = 32.7 and 127.7 µM, respectively)
Catalog No. 50-294-3821