Learn More
MSC PFERD Broom Head, 36 Inch, Coarse Sweep, Maroon Synthetic Fill, 3 Inch Trim
Supplier:  MSC 49167257
Encompass
This broom head is designed for heavy-duty sweeping of concrete and other rough surfaces. It is constructed from painted hardwood with two threaded holes. The water-resistant brown plastic fibers can be used wet or dry, are oil-resistant, and easy to clean, making it ideal for brushing construction rubble like gravel and grit.
  • Designed for heavy-duty sweeping of concrete and rough surfaces
  • Constructed from painted hardwood
  • Features two threaded holes
  • Water-resistant brown plastic fibers for wet or dry use
  • Oil-resistant and easy to clean
  • Ideal for brushing construction rubble, gravel, and grit
  • Overall length of 36 inches
  • Trim length of 3 inches
Catalog No. 50-283-7731