Learn More
ATCC Quantitative Synthetic Treponema pallidum DNA (ATCC BAA-2642SD™)
Supplier:  ATCC BAA2642SD
Encompass
Quantitative synthetic Treponema pallidum DNA (ATCC BAA-2642SD™) is a molecular standard used for assay development, validation, and qPCR calibration. It includes fragments from polA, 23S gene, 16S gene, flaA, 47kDa protein gene, and bmp. This product is manufactured under ISO 13485 guidance and supports infectious disease and sexually transmitted infection research.
Catalog No. 50-238-4034