Learn More
Cayman Chemical SynthetIc TropInon 5g
SDP
Supplier:  Cayman Chemical 342875 G
Encompass_Preferred
A tropane; a central intermediate in the biosynthesis of various tropane alkaloids
Catalog No. 50-294-9721