Learn More
Cayman Chemical SynthetIc Tyrosol 100g
SDP
Supplier:  Cayman Chemical 27600100 G
Encompass_Preferred
A phenol with diverse biological activities; fungicidal against H. capsulatum and C. posadasii (MFCs = 170-710 and 1,420 μg/ml, respectively); increases survival and pulmonary SOD activity and decreases pulmonary macrophage and neutrophil infiltration, edema, inflammation, and MPO activity in a mouse model of LPS-induced acute lung injury
Catalog No. 50-294-1651