Learn More
Sigma Aldrich Fine Chemicals Biosciences SynthwareTM all glass vacu1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SYNM2800051EA
Encompass
With 4 mm double oblique high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Catalog No. 50-290-8733