Learn More
Bachem Syntide 2 Trifluoroacetate
SDP
Supplier:  Bachem 4016901.0005
Encompass_Preferred
Syntide 2 Trifluoroacetate, H-Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-Gly-Leu-Pro-Gly-Lys-Lys-OH, 108334-68-5, C68H122N20O18, Mr 1507.84, 5mg. Calmodulin-Dependent Protein Kinase Substrate. Ca2+ calmodulin-dependent protein kinase II and Ca2+ phospholipid-dependent protein kinase (PKC) were selectively assayed using syntide 2 as substrate. The sequence of syntide 2 is homologous to phosphorylation site 2 in glycogen synthase.
Catalog No. 50-260-239