Learn More
Medchemexpress LLC Syntide 2 TFA | 108334-68-5 | 99.3% | 1621.84 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP0271A10MG
Encompass_Preferred
Syntide 2 TFA is a Ca2+- and calmodulin (CaM)-dependent protein kinase II (CaMKII) substrate peptide.
  • Selectively inhibits the gibberellin (GA) response
  • Leaves constitutive and abscisic acid-regulated events unaffected
Catalog No. 50-004-25227