Learn More
Medchemexpress LLC α-Synuclein inhibitor 10 | 3050715-65-3 | 99.9% | 409.78 g·mol⁻¹ | C21H12ClNO6 | 10 MG

Supplier: Medchemexpress LLC HY15730810MG
α-Synuclein inhibitor 10 is a small-molecule research compound that inhibits alpha-synuclein aggregation and is intended for use in Parkinson's disease and neurodegeneration studies.
- Inhibits α-synuclein aggregation; suitable for PD research.
- High purity: 99.85%.
- Solid form, light yellow to yellow.
- Molecular weight 409.78 g·mol⁻¹; formula C21H12ClNO6.
- Soluble in DMSO at approximately 25 mg/mL; use ultrasonic warming and optional heating to 60°C.
- Store protected from light; in solvent: -80°C for 6 months, -20°C for 1 month.
- COA and SDS available for quality and safety verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.