Learn More
Trajan Scientific L-MARK Brand 10l Syringe For Agilent, FN, 23g, 42mm, Pt. AS, 3 Pack
Supplier:  Trajan Scientific LMK.30301363P
Encompass_Preferred
L-MARK Brand Syringe, For Agilent 7673, 7683, 7693A, 6850 AutoSamplers, Similar to Agilent Part Number 9301-0713, 10uL, Fixed Needle (FN), 23g, 42mm, Point Style AS, Fitted Plunger, 3 Pack Made in Germany.
Catalog No. 50-125-1228